Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Peroxiredoxin 1 Mouse Recombinant Protein
SDP

Catalog No. NBP2523401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2523401 0.01 mg
NBP25234005 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2523401 Supplier Novus Biologicals™ Supplier No. NBP2523400.01MG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-199 of Mouse Peroxiredoxin 1 The Recombinant Mouse Peroxiredoxin 1 Protein is derived from E. coli. The Recombinant Mouse Peroxiredoxin 1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 5052
Molecular Weight (g/mol) TMW: 23.2kDa
Quantity 0.01 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias EC 1.11.1, EC 1.11.1.15, Natural killer cell-enhancing factor A, natural killer-enhancing factor A, NKEFA, NKEF-A, PAG, PAGAMSP23, PAGB, peroxiredoxin 1, peroxiredoxin-1, proliferation-associated gene A, Proliferation-associated gene protein, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2, Thioredoxin-dependent peroxide reductase 2
Common Name Peroxiredoxin 1
Biological Activity Specific activity is >2,500 pmol/min/ug. Enzymatic activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for minute.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.