Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07609IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-609-IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-609-IMI Supplier MilliporeSigma Supplier No. 07609I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Peroxiredoxin 1 Antibody is a highly specific rabbit polyclonal antibody that targets Peroxiredoxin 1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 1
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PRDX1,PAGA,PAGB,TDPX2
Host Species Rabbit
Immunogen KLH-conjugated linnear peptide corresponding to 12 amino acids from the C-terminal region of human Peroxiredoxin-1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_002565
Test Specificity This polyclonal antibody detects Peroxiredoxin-1 by Western blotting and immunocytochemistry in in DU145 cells and HeLa cells, respectively.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.