Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin 2 Antibody (SAIC-34D-11), DyLight 488 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320103G
Description
Peroxiredoxin 2 Monoclonal antibody specifically detects Peroxiredoxin 2 in Human samples. It is validated for Western BlotSpecifications
Peroxiredoxin 2 | |
Monoclonal | |
DyLight 488 | |
EC 1.11.1, MGC4104, Natural killer cell-enhancing factor B, natural killer-enhancing factor B, NKEFBNKEF-B, peroxiredoxin 2, peroxiredoxin-2, PRPTDPX1, PRX2, PRXII, thiol-specific antioxidant 1, Thiol-specific antioxidant protein, Thioredoxin peroxidase 1, Thioredoxin-dependent peroxide reductase 1, torin, TPX1, TSAEC 1.11.1.15 | |
Peptide "e;DHPFGFVAVPTK"e;, derived from the cellular redox regulator Peroxiredoxin-2, conjugated to KLH. | |
0.1 mL | |
Cancer | |
7001 | |
Store at 4&drg;C in the dark. | |
IgG κ |
Western Blot | |
SAIC-34D-11 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction