Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
peroxiredoxin 2 Mouse anti-Human, Mouse, Clone: 3H6C4, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 602021IG150UL

Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. The crystal structure of this protein has been resolved to 2.7 angstroms. Transcript variants encoding distinct isoforms have been identified for this gene.Specifications
peroxiredoxin 2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32119, Q61171 | |
PRDX2 | |
peroxiredoxin 2 Fusion Protein Ag0835 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3H6C4 | |
Unconjugated | |
PRDX2 | |
NKEF B, NKEFB, peroxiredoxin 2, PRDX2, PRP, PRX2, PRXII, TDPX1, Thioredoxin peroxidase 1, TSA | |
Mouse | |
Protein G | |
RUO | |
21672, 7001 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction