Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin 3 Antibody (AT1F8), Alexa Fluor™ 488 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP312869AF488
Description
Peroxiredoxin 3 Monoclonal antibody specifically detects Peroxiredoxin 3 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Peroxiredoxin 3 | |
Monoclonal | |
Alexa Fluor 488 | |
Antioxidant protein 1MGC104387, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial | |
Recombinant human Peroxiredoxin3 (63-256aa) purified from E. coli (Accession number: NP_006784) | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
10935 | |
Store at 4°C in the dark. | |
IgG3 κ |
Western Blot, ELISA, Immunofluorescence | |
AT1F8 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction