Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin 4 Antibody (SAIC-40C-8), Alexa Fluor™ 594 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320106AF594
Description
Peroxiredoxin 4 Monoclonal antibody specifically detects Peroxiredoxin 4 in Human samples. It is validated for Western Blot, ELISASpecifications
Peroxiredoxin 4 | |
Monoclonal | |
Alexa Fluor 594 | |
Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372 | |
Peptide "e;LVQAFQYTDK"e; derived from the cell redox-regulator Peroxiredoxin-4, conjugated to KLH. | |
0.1 mL | |
Signal Transduction | |
10549 | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, ELISA | |
SAIC-40C-8 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction