Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin 5 Antibody (002), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289862H
Description
Peroxiredoxin 5 Monoclonal specifically detects Peroxiredoxin 5 in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
Peroxiredoxin 5 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
25824 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunoassay | |
2 | |
Western Blot, Knockout Validated | |
ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Peroxiredoxin 5 (P30044-2; Met53-Leu214). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction