Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 6 (PRDX6) Monoclonal Antibody (OTI11B8), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene CF813658

Encompass
Encompass_Preferred

Catalog No. 50-254-3652


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Peroxiredoxin 6 (PRDX6) Monoclonal antibody specifically detects Peroxiredoxin 6 (PRDX6) in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Peroxiredoxin 6 (PRDX6)
Monoclonal
See Label
PBS with 8% trehalose and no preservative; pH 7.3
O08709, O35244, P30041
Mouse
Protein A/G
RUO
11758, 94167, 9588
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI11B8
Unconjugated
PRDX6
1-Cys; 1-Cys peroxiredoxin; 1-Cys PRX; 1-cysPrx; 24 kDa protein; 9430088D19Rik; AA690119; Acidic calcium-independent phospholipase A2; aiPLA2; Antioxidant protein 2; Aop2; Brp-12; CP-3; epididymis secretory sperm binding protein Li 128m; GPx; HEL-S-128m; KIAA0106; Liver 2D page spot 40; Ltw4; Ltw-4; Lvtw-4; MGC46173; MSP23; NKEFA; Non-selenium glutathione peroxidase; NSGPx; ORF06; p29; PAG; PAGA; PAGB; peroxiredoxin 6; peroxiredoxin-6; Prdx5; Prdx6; PRX; Prx6; Red blood cells page spot 12; thiol-specific antioxidant protein; Tsa
Full length human recombinant protein of human Prdx6 (NP_004896) produced in E.coli.
100 μg
Primary
Human, Mouse, Rat
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.