Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin, bacteria-like protein Antibody (V71), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259672F
Description
Peroxiredoxin, bacteria-like protein Monoclonal specifically detects Peroxiredoxin, bacteria-like protein in Parasite samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Peroxiredoxin, bacteria-like protein | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Peptide Sequence specific to peroxiredoxin antigen from Tetracapsuloides bryosalmonae (fish parasite). | |
0.1 mL | |
Virology Bacteria and Parasites | |
9682623 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
V71 | |
ELISA, Immunohistochemistry | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Parasite | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction