Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PERP Recombinant Rabbit Monoclonal Antibody (1O3L5)
GREENER_CHOICE

Catalog No. PIMA538148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538148 Supplier Invitrogen™ Supplier No. MA538148
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HepG2, BxPC-3, A-431, Mouse liver, Mouse kidney, Rat lung, Rat liver Immunogen sequence: LSFFALCGPQ MLVFLRVIGG LLALAAVFQI ISLVIYPVKY TQTFTLHANP AVTYIYNWAY GFGWAATIIL IGCAFFFCCL PNYEDDLLGN AKPRYFYTSA.

The p53 tumor-suppressor gene integrates numerous signals that control cell life and death. Several novel molecules involved in p53 network, including Chk2, p53R2, p53AIP1, Noxa, PIDD, PID/MTA2, MTBP and PERP, were identified and their genes were cloned recently. PERP, also termed PIGPC1 and THW, is a plasma membrane protein. p53 binds to the promoter of PERP and transcriptionally activates PERP gene then the translated PERP protein mediates the p53 induced apoptosis. The expression of PERP causes cell death. PERP is a mediator of p53 induced apoptosis. PERP has sequence similarity to PMP-22/gas3 and is a new member of the PMP-22/gas3 family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PERP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1O3L5
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PERP
Gene Accession No. Q96FX8, Q9JK95
Gene Alias 1110017A08Rik; dJ496H19.1; HGNC:17637; KCP1; KCP-1; keratinocyte associated protein 1; keratinocyte-associated protein 1; keratinocytes associated protein 1; KRTCAP1; p53 apoptosis effector related to PMP22; p53 apoptosis effector related to PMP-22; p53 apoptosis-associated target; p53-induced protein PIGPC1; PERP; PERP, TP53 apoptosis effector; PIGPC1; RP3-496H19.1; THW; Transmembrane protein THW
Gene Symbols PERP
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292949, 64058, 64065
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.