Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PERQ2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212813AF532
Description
PERQ2 Polyclonal antibody specifically detects PERQ2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PERQ2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
DKFZp686J17223, FLJ23368, GRB10 interacting GYF protein 2, GRB10-interacting GYF protein 2, GYF domain containing 2, GYF2, KIAA0642DKFZp686I15154, PARK11, Parkinson disease (autosomal recessive, early onset) 11, PERQ amino acid rich, with GYF domain 2, PERQ amino acid rich, with GYF domain 3, PERQ amino acid-rich with GYF domain-containing protein 2, PERQ2, PERQ3, TNRC15, trinucleotide repeat containing 15, Trinucleotide repeat-containing gene 15 protein | |
The immunogen this antibody was made to, maps to a region between residue 1249 and 1299 of human GRB10 Interacting GYF protein 2 using the numbering given in entry NP_001096616.1 (GeneID 26058). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
26058 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction