Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pertussis Toxin
GSA_VAGREENER_CHOICESDP

Catalog No. 309750U Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-975-0U 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-975-0U Supplier Tocris Bioscience™ Supplier No. 3097/50U
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Catalyzes ADP-ribosylation of Gi, Go and Gt

Pertussis Toxin is a bacterial toxin that catalyzes ADP-ribosylation of G-proteins Gi, Go and Gt. Impairs G protein heterotrimer interaction with receptors, blocking receptor coupling. Pertussis Toxin protects against ischemic stroke in a mouse model of permanent middle cerebral artery occlusion. Pertussis Toxin inhibits microglia migration to the spinal cord and prevents dissemination of disease in an experimental autoimmune encephalomyelitis mouse model.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 70323-44-3

Specifications

Quantity 50 μg
Purity >98%
Chemical Name or Material Pertussis Toxin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.