Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PES1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583491
Description
Immunogen sequence: RMEGKKPRVM AGTLKLEDKQ RLAQEEESEA KRLAIMMMKK REKYLYQKIM FGKRRKIREA NKLAEKRKAH DE.
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
Specifications
PES1 | |
Polyclonal | |
Unconjugated | |
Pes1 | |
HAMAP-Rule:MF_03028}; Pes; PES1; Pescadillo homolog; pescadillo homolog {ECO:0000255; pescadillo homolog 1, containing BRCT domain; pescadillo homolog 1, containing BRCT domain (zebrafish); pescadillo ribosomal biogenesis factor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23481 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00541 | |
Pes1 | |
Recombinant protein corresponding to Human PES1. Recombinant protein control fragment (Product #RP-104715). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction