Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PES1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584733 Supplier Invitrogen™ Supplier No. PA584733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYQLLNLHYP PKLEGQAQAE AKAGEGTYAL DSESCMEKLA ALSASLARVV VPATEEEAEV DEFPTDGEMS AQEEDRRKE Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 84%.

Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PES1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pes1
Gene Accession No. O00541
Gene Alias HAMAP-Rule:MF_03028}; Pes; PES1; Pescadillo homolog; pescadillo homolog {ECO:0000255; pescadillo homolog 1, containing BRCT domain; pescadillo homolog 1, containing BRCT domain (zebrafish); pescadillo ribosomal biogenesis factor 1
Gene Symbols Pes1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PES1. Recombinant protein control fragment (Product #RP-105400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23481
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.