Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Aldrin Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. p-7187794
Click to view available options
Quantity:
2 mL
10 mL

Aldrin belongs to the class of organochlorine pesticides, commonly used in agriculture and is persistent as a highly toxic environmental contaminant.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 309-00-2
Linear Formula C12H8 Cl6
MDL Number MFCD00135587
Type Analytical Standard
Quantity 10 mL
Formula Weight 364.91
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.