Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PESTANAL™ Chloramben, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
100 mg
Description
Agriculture

Specifications
Specifications
Product Line | PESTANAL |
CAS | 133-90-4 |
Physical Form | Neat |
Linear Formula | H2 NC6H2(Cl)2 CO2 H |
MDL Number | MFCD00065093 |
Melting Point | 194°C to 197°C |
Type | Analytical Standard |
Quantity | 100 mg |
Formula Weight | 206.03 |
Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction