Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PESTANAL™ Malathion Solution, 100 μg/mL in cycloHexane, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
2 mL
10 mL
Description
Agriculture

Specifications
Specifications
Product Line | PESTANAL |
CAS | 121-75-5 |
Flash Point | -18°C - Closed Cup |
Linear Formula | C10H19O6 PS2 |
MDL Number | MFCD00036233 |
Type | Analytical Standard |
Quantity | 10 mL |
Formula Weight | 330.36 |
Grade | Analytical Standard |
Concentration or Composition (by Analyte or Components) | 100 μg/mL in cyclohexane |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction