Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Nitrobenzene, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187591
Click to view available options
Quantity:
1 mL
5 mL

Nitrobenzene is a carcinogenic compound which has been employed extensively in dyes, pesticides and explosives industries.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 98-95-3
Product Line PESTANAL
Ignition Point 899°F
Vapor Pressure 0.15 mmHg (20°C)
Vapor Density 4.2 (vs. air)
Density 1.196 g/mL (at 25°C)
Flash Point 88°C
Linear Formula C6H5 NO2
MDL Number MFCD00007043
Melting Point 5°C to 6°C
Boiling Point 210°C to 211°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 123.11
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.