Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Phenol Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. p-7187733 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
2 mL
10 mL

Phenol belongs to the class of aromatic volatile organic compounds (VOCs).

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 108-95-2
Flash Point 2°C
Linear Formula C6H6 O
MDL Number MFCD00002143
Type Analytical Standard
Quantity 2 mL
Formula Weight 94.11
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.