Learn More
Restek™ Pesticide Residue Cleanup SPE Cartridges
SDP
Catalog No. 06729304 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Base Material:
200 mg CarboPrep 200/400 mg PSA
500 mg CarboPrep 200/500 mg PSA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Base Material
06-729-304 200 mg CarboPrep 200/400 mg PSA
06-729-306 500 mg CarboPrep 200/500 mg PSA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-729-304 Supplier Restek™ Supplier No. 26127
Add to Cart
Add to Cart
Convenient, multiple adsorbent beds in a single cartridge.
  • For use in multiresidue pesticide analysis to remove matrix interferences
  • Excellent for cleanup of dietary supplement extracts
  • 6mL Combo SPE Cartridge
  • Packed with 200mg CarboPrep 200 and 400mg PSA, PTFE frits
  • Quantity: 30/Pk.
Specifications
Base Material 200 mg CarboPrep 200/400 mg PSA
For Use With (Application) Multiresidue Pesticide Analysis to Remove Matrix Interferences, Cleanup of Dietary Supplement Extracts
Type 6 mL Combo SPE Cartridge