Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PET100 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584753 Supplier Invitrogen™ Supplier No. PA584753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELWPPEKLQE IEEFKERLRK RREEKLLRDA QQ Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 60%.

Mitochondrial complex IV, or cytochrome c oxidase, is a large transmembrane protein complex that is part of the respiratory electron transport chain of mitochondria. The small protein encoded by this gene plays a role in the biogenesis of mitochondrial complex IV. This protein localizes to the inner mitochondrial membrane and is exposed to the intermembrane space. Mutations in this gene are associated with mitochondrial complex IV deficiency. This gene has a pseudogene on chromosome 3. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Apr 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PET100
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PET100
Gene Accession No. P0DJ07
Gene Alias C19orf79; PET100; PET100 homolog; protein PET100 homolog, mitochondrial
Gene Symbols PET100
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PET100. Recombinant protein control fragment (Product #RP-108525).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.