Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PET117 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297948C
Description
PET117 Polyclonal specifically detects PET117 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PET117 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
PET117 PET117 homolog (S. cerevisiae) | |
Produced in rabbits immunized with E. coli-derived Human PET117 fragment. | |
0.1 mL | |
metabolism | |
100303755 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction