Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ Petri Dish Platinum Electrode for Tissues Chamber Kit

Catalog No. p-9571002 Shop All BTX Harvard Products
Change view
Click to view available options
Size (Metric) Gap:
5 mm
15 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Gap
BTX450505 5 mm
BTX450507 15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX450505 Supplier BTX™ Supplier No. 450505
Only null left
Add to Cart
Add to Cart

Petri Dish Platinum Electrode for Tissues Chamber Kit electrodes and chambers for ex vivo tissues gene or drug delivery.

  • Simple and easy to use
  • Available in 5 and 15 mm
  • Reusable, lab-grade Pyrex glass and two platinum electrodes

Specifications

For Use With (Application) For ex Vivo Tissues Gene or Drug Delivery Using Large or Uniquely Shapped Tissue Samples
For Use With (Equipment) ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001
Material Platinum
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 200 V
Description Petri Dish Platinum Electrode for Tissues Chamber Kit, 5 mm gap
Depth (English) 0.12 in.
Depth (Metric) 3 mm
Includes Petri Dish Electrode Chamber and Electrodes
Length (English) 0.31 in.
Length (Metric) 8 mm
Pulse Length Range 10 μsec. to 100 msec.
Quantity 1 Ea.
Size (English) Gap 0.2 in.
Size (Metric) Gap 5 mm
Width (English) 0.2 in.
Width (Metric) 5 mm
Sterility Non-sterile
Connection Type Banana Plug to Micrograbber
Pulse Form Square Wave
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.