Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetamide, 99+%, ACS reagent

Catalog No. AC424531000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 62-55-5 | C2H5NS | 75.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-55-5
Molecular Formula C2H5NS
Molecular Weight (g/mol) 75.13
MDL Number MFCD00008070
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
PubChem CID 2723949
ChEBI CHEBI:32497
IUPAC Name ethanethioamide
SMILES CC(N)=S

Specifications

Melting Point 112.0°C to 114.0°C
Color White to Beige
Density 1.3700g/mL
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3CSNH2
Quantity 100 g
Beilstein 02, 232
Fieser 04,502
Specific Gravity 1.37
Merck Index 15, 9472
Solubility Information Solubility in water: 163g/L (25°C). Other solubilities: 26.4g/100g ethanol, sparingly soluble in ether, soluble in benzene
Formula Weight 75.13
Percent Purity 99+%
Clarity (2% soln.) Passes test
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material Thioacetamide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water
EINECSNumber 200-541-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.