Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether 35/60, Environmental Grade

Catalog No. p-4408282
Click to view available options
Quantity:
1 L
4 L

Suitable For Environmental and Pesticide Analysis | CAS: 8032-32-4

It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Color Colorless
Appearance Colorless liquid
Density 0.64
Boiling Point 35°C to 60°C
Quantity 1 L
Flash Point -49°C (-57°F)
Ignition Point 250°C (482°F)
Odor Petroleum-like
Physical Form Liquid
Refractive Index 1.363
Assay Percent Range >98%
Vapor Pressure 350 hPa (263mm Hg) at 20°C
Solubility Information Insoluble in water.
Grade Environmental
Chemical Name or Material Petroleum ether 35/60, Environmental Grade
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packaging Group: II
EINECSNumber 232-453-7
RTECSNumber OI6180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.