Learn More
Petroleum Ether, ACS Reagent (Ligroin), Reagents
Specifications
Specifications
| Chemical Name or Material | Petroleum Ether, ACS Reagent (Ligroin) |
| CAS | 8032-32-4 |
| Color | Colorless |
| Boiling Point | 35°C |
| Quantity | 4 L |
| Packaging | Safety Coated Glass |
| Physical Form | Liquid |
| Specific Gravity | 0.645 |
| Molecular Formula | C6-16H12-34 |
| Synonym | Naphtha, Petroleum benzine, Petroleum spirit, Ligroin, Ligroine |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.