Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Petroleum Ether, ACS Reagent (Ligroin), Reagents
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Glass Bottle
Safety Coated Glass
Steel Drum
Steel Pail
Tin Can
Specifications
Specifications
| Chemical Name or Material | Petroleum Ether, ACS Reagent (Ligroin) |
| CAS | 8032-32-4 |
| Color | Colorless |
| Boiling Point | 35°C |
| Quantity | 200 L |
| Packaging | Steel Drum |
| Physical Form | Liquid |
| Specific Gravity | 0.645 |
| Molecular Formula | C6-16H12-34 |
| Synonym | Naphtha, Petroleum benzine, Petroleum spirit, Ligroin, Ligroine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction