Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Petroleum Ether/Dichloromethane, 80/20 v/v, Reagents
Click to view available options
Quantity:
2.5 L
4 L
Specifications
Specifications
Color | Colorless |
Quantity | 4 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Specific Gravity | 0.7813 |
Grade | Reagent |
Chemical Name or Material | Petroleum Ether/Dichloromethane, 80/20 v/v |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction