Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpyrrolidone, Honeywell Burdick & Jackson™

Catalog No. 60090422 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
20 L
Packaging:
Returnable NOWPAK™ II
This item is not returnable. View return policy

Catalog No. 60-090-422

Supplier: Honeywell Chemicals BN30520

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

BioSyn™, for biosynthesis and applications requiring low water content, >99.5%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
PubChem CID 13387
ChEBI CHEBI:7307

Specifications

Quantity 20 L
Flash Point 88 °C
Packaging Returnable NOWPAK™ II
Physical Form Liquid, Clear
Refractive Index 1.4680 - 1.4720
Grade Low Water
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.