Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, puriss. p.a., Honeywell™ Riedel-de Haën™

Catalog No. 60048554 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
2.5 L
Packaging:
Stainless Steel Can

Catalog No. 60-048-554

Supplier: Honeywell Chemicals 773992.5L

Only null left
Add to Cart
Add to Cart

Puriss. p.a., bp 30-40 °C, low boiling point hydrogen treated naphtha

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0, 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
IUPAC Name pentane
SMILES CCCCC

Specifications

Density 0.63g/mL at 20°C
Boiling Point 30°C to 40°C
Quantity 2.5 L
Flash Point −49°C (−56.2°F)
Packaging Stainless Steel Can
UN Number UN1268
Grade puriss. p.a.
Chemical Name or Material Petroleum ether
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
DOTInformation class 3 - PG 1 - EHS - Petroleum distillates, n.o.s., HI: S
RTECSNumber 309-852-0
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.