Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Petroleum Ether, puriss., Honeywell™ Riedel-de Haën™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
5 L
4 x 2.5 L
4 x 5 L
Packaging:
Aluminum Bottle
Glass Bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.

Specifications
Specifications
Density | 0.642 to 0.656g/mL at 20°C |
Quantity | 4 x 2.5 L |
Flash Point | −40°C (−40°F) |
Packaging | Glass Bottle |
UN Number | UN1268 |
Grade | puriss. |
Chemical Name or Material | Petroleum Ether, Meets analytical specification of DAB |
Safety and Handling
Hazard Statement | P210-P273-P301 + P310-P331-P391-P403 + P235 |
Precautionary Statement | H225-H304-H336-H411 |
DOTInformation | Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S |
EINECSNumber | 309-852-0 |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction