Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
195 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications

Specifications
Specifications
Appearance | Liquid |
Density | 0.640 to 0.655g/mL at 20°C |
Boiling Point | 40°C to 60°C |
Quantity | 2.5 L |
Flash Point | −30°C (−22°F) |
Packaging | Glass Bottle |
Residue after Evaporation | 0 to 0.001% |
Titratable Acid | Free acid (as CH3COOH): ≤0.002% |
UN Number | UN1268 |
Grade | ACS/puriss. p.a. |
Show More |
Safety and Handling
Hazard Statement | P210-P261-P273-P301 + P310-P331 |
Precautionary Statement | H225-H302-H304-H315-H336-H411 |
DOTInformation | Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction