Learn More
Justrite PetroPad
Catalog No. 17988513 Shop All Justrite Safety Group Products
Click to view available options
Dimensions (L x W):
24 x 18.1 in.
35.4 x 27.2 in.
53.9 x 35.8 in.
53.9 x 53.9 in.
85.8 x 53.9 in.
Reusable, washable and easy to dispose absorbent pad designed for the safe handling and the disposal of hazardous hydrocarbon waste.
  • Portable and easy to fold up and store without fear of leakage or mess
  • Absorbs and permanently solidifies hydrocarbons while allowing rainwater to pass through
  • Durable material and robust construction withstand the demands of a rigorous work environment
  • Easily pressure wash built-up dirt and debris without releasing hydrocarbons
  • TCLP leachate test and EPA Paint Filter Test (9095B) compliant
Specifications
Material Polymer
Type Sorbent, Oil-only
Disposable Reusable
Weight Capacity Heavyweight
Dimensions (L x W) 53.9 x 53.9 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.