Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145449
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145449 200 μL
FEPA5145450 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145449 Supplier Invitrogen™ Supplier No. PA5145449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the AAA ATPase family, a large group of ATPases associated with diverse cellular activities. This protein is cytoplasmic but is often anchored to a peroxisomal membrane where it forms a heteromeric complex and plays a role in the import of proteins into peroxisomes and peroxisome biogenesis. Mutations in this gene have been associated with complementation group 1 peroxisomal disorders such as neonatal adrenoleukodystrophy, infantile Refsum disease, and Zellweger syndrome. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Pex1
Gene Accession No. O43933, Q5BL07
Gene Alias 5430414H02Rik; E330005K07Rik; HMLR1; PBD1A; PBD1B; Peroxin-1; peroxisomal biogenesis factor 1; peroxisome biogenesis disorder protein 1; Peroxisome biogenesis factor 1; Pex1; RGD1559939; Zellweger syndrome; ZWS; ZWS1
Gene Symbols Pex1
Host Species Rabbit
Immunogen Recombinant Human PEX1 protein, Glu62-Gln219 (Accession #O43933).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100626142, 5189, 71382
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.