Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEX10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562034
Description
Immunogen sequence: QRARKEWRLH RGLSHRRASL EERAVSRNPL CTLCLEERRH PTATPCGHLF CWECITAWCS SKAECPLCRE KFPPQK Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
This gene encodes a protein involved in import of peroxisomal matrix proteins. This protein localizes to the peroxisomal membrane. Mutations in this gene result in phenotypes within the Zellweger spectrum of peroxisomal biogenesis disorders, ranging from neonatal adrenoleukodystrophy to Zellweger syndrome. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].
Specifications
PEX10 | |
Polyclonal | |
Unconjugated | |
PEX10 | |
AV128229; Gm142; MGC1998; NALD; PBD6A; PBD6B; peroxin 10; peroxin-10; Peroxisomal biogenesis factor 10; peroxisome assembly protein 10; Peroxisome biogenesis factor 10; Pex10; RING finger protein 69; RNF69 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5192 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60683 | |
PEX10 | |
Recombinant protein corresponding to Human PEX10. Recombinant protein control fragment (Product #RP-98534). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction