Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103999
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103999 Supplier Invitrogen™ Supplier No. PA5103999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PEX11A.

PEX11A may be involved in peroxisomal proliferation and may regulate peroxisomes division. It may mediate binding of coatomer proteins to the peroxisomal membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX11A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PEX11A
Gene Accession No. O70597, O75192, Q9Z211
Gene Alias 28 kDa peroxisomal integral membrane protein; hsPEX11p; Peroxin-11A; peroxisomal biogenesis factor 11 alpha; peroxisomal biogenesis factor 11a; Peroxisomal coatomer receptor; peroxisomal membrane protein 11A; Peroxisomal membrane protein 26; peroxisomal membrane protein Pmp26p (Peroxin-11); Pex11; Pex11a; PEX11alpha; PEX11-alpha; pex11palpha; Pmp26p; PMP28; protein PEX11 homolog alpha; RnPEX11p
Gene Symbols PEX11A
Host Species Rabbit
Immunogen A synthesized peptide derived from human PEX11A(Accession O75192), corresponding to amino acid residues L132-L182.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18631, 85249, 8800
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.