Learn More
Invitrogen™ PEX11A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113818
Description
Immunogen sequence: MKRVTCDRAK KEKSASQDPL WFSVAEEETE WLQSFLLLLF RSLKQHPPLL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 79%; Human: 100%; Pig: 77%.
PEX11A may be involved in peroxisomal proliferation and may regulate peroxisomes division. It may mediate binding of coatomer proteins to the peroxisomal membrane.
Specifications
PEX11A | |
Polyclonal | |
Unconjugated | |
PEX11A | |
28 kDa peroxisomal integral membrane protein; hsPEX11p; Peroxin-11A; peroxisomal biogenesis factor 11 alpha; peroxisomal biogenesis factor 11a; Peroxisomal coatomer receptor; peroxisomal membrane protein 11A; Peroxisomal membrane protein 26; peroxisomal membrane protein Pmp26p (Peroxin-11); Pex11; Pex11a; PEX11alpha; PEX11-alpha; pex11palpha; Pmp26p; PMP28; protein PEX11 homolog alpha; RnPEX11p | |
Rabbit | |
Affinity chromatography | |
RUO | |
8800 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75192 | |
PEX11A | |
Synthetic peptide directed towards the middle region of human PEX11A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.