Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEX11B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551034
Description
PEX11B / Peroxin 11B Polyclonal Antibody detects endogenous levels of PEX11B / Peroxin 11B protein.
Peroxisomal membrane protein 11B is a protein that in humans is encoded by the PEX11B gene. Involved in peroxisomal proliferation. May regulate peroxisomes division by recruiting the dynamin-related GTPase DNM1L to the peroxisomal membrane.
Specifications
PEX11B | |
Polyclonal | |
Unconjugated | |
PEX11B | |
Peroxin 11 beta; Peroxin 11B; Peroxin-11 beta; Peroxin11B; peroxin-11B; peroxisomal biogenesis factor 11 beta; peroxisomal biogenesis factor 11B; Peroxisomal membrane protein 11B; PEX11 beta; PEX-11 beta; PEX11B; PEX11beta; PEX11-beta; Pex11pbeta; PEX14B; Protein PEX11 homolog beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18632, 310682, 8799 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O96011, Q9Z210 | |
PEX11B | |
Synthesized peptide derived from the Internal region of human Peroxin 11β. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction