Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX11G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539331
Description
The protein encoded by this gene is a member of the PEX11 family. This family is reported to regulate the number and size of peroxisomes in evolutionarily distant organisms. The protein encoded by this gene may induce clustering of peroxisomes. Alternative splicing results in multiple transcript variants that encode different protein isoforms. [provided by RefSeq, Jul 2012].
Specifications
PEX11G | |
Polyclonal | |
Unconjugated | |
PEX11G | |
PEX11C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92960 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96HA9 | |
PEX11G | |
A synthetic peptide derived from the N-terminal region of human PEX11C | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction