Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX11G Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2833830.1ML
Description
PEX11G Polyclonal specifically detects PEX11G in Human samples. It is validated for Western Blot.Specifications
PEX11G | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
92960 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
MGC4281, peroxin Pex11p gamma, Peroxin-11C, peroxisomal biogenesis factor 11 gamma, Peroxisomal biogenesis factor 11C, peroxisomal biogenesis factor 11G, peroxisomal membrane protein 11C, PEX11C, PEX11-gamma, Protein PEX11 homolog gamma | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human PEX11G | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction