Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564414 Supplier Invitrogen™ Supplier No. PA564414
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAQHHSPLLR LAGVQLGRLT VQDIQALEHK PAKASMMQQP ARSVSEKINS AL Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 69%.

Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEX12
Gene Accession No. O00623
Gene Alias AI451906; PAF3; PAF-3; PBD3A; peroxin 12; peroxin-12; peroxisomal biogenesis factor 12; peroxisome assembly factor 3; Peroxisome assembly protein 12; Pex12
Gene Symbols PEX12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEX12. Recombinant protein control fragment (Product #RP-103796).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.