Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556942 Supplier Invitrogen™ Supplier No. PA556942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SADLGPTLMT RPGQPALTRV PPPILPRPSQ QTGSSSVNTF RPAYSSFSSG YGAYGNSFYG GYSPYSYGYN GLGYNRLRVD DLPPSRFVQQ AEESSRGA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

PEX13 (Peroxisomal Biogenesis Factor 13) is a gene located on chromosome 2p15 that encodes a peroxisomal membrane protein essential for the biogenesis and function of peroxisomes, which are cellular organelles involved in lipid metabolism and detoxification of reactive oxygen species. PEX13 is a component of the peroxisomal import machinery that specifically recognizes and binds to peroxisomal targeting signal 1 (PTS1) containing proteins, facilitating their import into the peroxisome matrix. Mutations in the PEX13 gene can lead to Zellweger spectrum disorders, a group of peroxisome biogenesis disorders (PBDs) characterized by defects in multiple peroxisomal metabolic pathways, leading to severe developmental issues, neurological deficits, and often early lethality. Studies of PEX13 have helped elucidate the mechanisms of peroxisomal protein import and the broader implications of peroxisomal dysfunction in human disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEX13
Gene Accession No. Q92968
Gene Alias NALD; PBD11A; PBD11B; Peroxin-13; peroxisomal biogenesis factor 13; peroxisomal membrane protein PEX13; peroxisome biogenesis factor 13; Pex13; ZWS
Gene Symbols PEX13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEX13. Recombinant protein control fragment (Product #RP-95803).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5194
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.