Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX14 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171841F
Description
PEX14 Polyclonal antibody specifically detects PEX14 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
PEX14 | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
dJ734G22.2, MGC12767, NAPP2, NF-E2 associated polypeptide 2, peroxin-14, peroxisomal biogenesis factor 14, Peroxisomal membrane anchor protein PEX14, peroxisomal membrane anchor protein Pex14p, peroxisomal membrane protein PEX14, Pex14p, PTS1 receptor docking protein, PTS1 receptor-docking protein | |
The immunogen for this product maps to a region between residue 250 and 300 of human Peroxisomal Biogenesis Factor 14 using the numbering given in entry NP_004556.1 (GeneID 5195). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5195 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction