Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX14 (Human Specific) Rabbit anti-Human, Unconjugated, Clone: 6K5, Proteintech
SDP

Catalog No. 502256590 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6590 Supplier Proteintech Group Inc Supplier No. 801961RR100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Antibody

The PEX14 (human specific) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PEX14 (human specific). This antibody recognizes human antigen. The PEX14 (human specific) antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Antigen PEX14 (Human Specific)
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 6K5
Conjugate Unconjugated
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5195
Target Species Human
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.