Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561913 Supplier Invitrogen™ Supplier No. PA561913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAVNHHSSSD ISPVSNESTS SSPGKEGHSP EGSTVTYHLL GPQEEGEGVV DVKGQVRMEV QGEEEKREDK EDEE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

PEX14 belongs to the peroxin 14 family. It is a component of the peroxisomal translocation machinery with PEX13 and PEX17 and interacts with both the PTS1 and PTS2 receptors. PEX14 binds directly to PEX17. Defects in PEX14 are the cause of peroxisome biogenesis disorder complementation group K (PBD-CGK) and Zellweger syndrome (ZWS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX14
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEX14
Gene Accession No. O75381
Gene Alias dJ734G22.2; NAPP2; NF-E2 associated polypeptide 2; PBD13A; Peroxin 14; peroxin-14; peroxisomal biogenesis factor 14; peroxisomal membrane anchor protein PEX14; peroxisomal membrane anchor protein Pex14p; peroxisomal membrane protein PEX14; peroxisomal protein Pex14; Pex14; Pex14p; PEXE; PTS1 receptor docking protein; PTS1 receptor-docking protein; R75137
Gene Symbols PEX14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEX14. Recombinant protein control fragment (Product #RP-95797).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5195
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.