Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596642 Supplier Thermo Scientific Supplier No. PA596642
Add to Cart
Add to Cart

PEX14 Polyclonal Antibody

PEX14 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX14
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PEX14
Gene Accession No. O75381, Q9R0A0
Gene Alias dJ734G22.2; NAPP2; NF-E2 associated polypeptide 2; PBD13A; Peroxin 14; peroxin-14; peroxisomal biogenesis factor 14; peroxisomal membrane anchor protein PEX14; peroxisomal membrane anchor protein Pex14p; peroxisomal membrane protein PEX14; peroxisomal protein Pex14; Pex14; Pex14p; PEXE; PTS1 receptor docking protein; PTS1 receptor-docking protein; R75137
Gene Symbols PEX14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 118-377 of human PEX14 (NP_0045561).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5195, 56273
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.