Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX14 Recombinant Rabbit Monoclonal Antibody (7E6), Proteintech
SDP

Catalog No. 502256596 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6596 Supplier Proteintech Group Inc Supplier No. 802341RR100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PEX14 Recombinant Monoclonal antibody specifically detects PEX14 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PEX14
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene H3C1
Gene Accession No. P68431, P68433
Gene Alias dJ734G22.2; NAPP2; NF-E2 associated polypeptide 2; PBD13A; Peroxin 14; peroxin-14; peroxisomal biogenesis factor 14; peroxisomal membrane anchor protein PEX14; peroxisomal membrane anchor protein Pex14p; peroxisomal membrane protein PEX14; peroxisomal protein Pex14; Pex14; Pex14p; PEXE; PTS1 receptor docking protein; PTS1 receptor-docking protein; R75137
Host Species Rabbit
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360198, 8350
Content And Storage -20°C
Form Liquid
Isotype IgG
Target Molecule Human|Mouse|Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.