Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560311 Supplier Invitrogen™ Supplier No. PA560311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGLQTSPPIV PLDRETQAQP PDGDHSPGNH EQSYVGKRSN RVVRTLQNTP SLHSRHWGAP QQREGRQQQH HEELSATPTP LGLQETIAE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

PEX16 is required for peroxisome membrane biogenesis. It may play a role in early stages of peroxisome assembly and can recruit other peroxisomal proteins, such as PEX3 and PMP34, to de novo peroxisomes derived from the endoplasmic reticulum (ER). It may function as receptor for PEX3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEX16
Gene Accession No. Q91XC9, Q9Y5Y5
Gene Alias PBD8A; PBD8B; peroxin 16; peroxin-16; Peroxisomal biogenesis factor 16; peroxisomal membrane protein PEX16; peroxisome biogenesis factor 16; Pex16
Gene Symbols PEX16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEX16. Recombinant protein control fragment (Product #RP-97322).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18633, 9409
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.