Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEX16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106318
Description
Antibody detects endogenous levels of total PEX16.
PEX16 is required for peroxisome membrane biogenesis. It may play a role in early stages of peroxisome assembly and can recruit other peroxisomal proteins, such as PEX3 and PMP34, to de novo peroxisomes derived from the endoplasmic reticulum (ER). It may function as receptor for PEX3.
Specifications
| PEX16 | |
| Polyclonal | |
| Unconjugated | |
| PEX16 | |
| PBD8A; PBD8B; peroxin 16; peroxin-16; Peroxisomal biogenesis factor 16; peroxisomal membrane protein PEX16; peroxisome biogenesis factor 16; Pex16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18633, 311203, 9409 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91XC9, Q9Y5Y5 | |
| PEX16 | |
| A synthesized peptide derived from human PEX16(Accession Q9Y5Y5), corresponding to amino acid residues Q117-V167. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction