Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 148161AP150UL

Description
PEX16 is required for peroxisome membrane biogenesis. It may play a role in early stages of peroxisome assembly and can recruit other peroxisomal proteins, such as PEX3 and PMP34, to de novo peroxisomes derived from the endoplasmic reticulum (ER). It may function as receptor for PEX3.Specifications
PEX16 | |
Polyclonal | |
Unconjugated | |
PEX16 | |
Peroxin 16, PEX16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18633, 311203, 9409 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91XC9, Q9Y5Y5 | |
Pex16 | |
PEX16 Fusion Protein Ag6566 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction